Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Test Tube Racks, Globe Scientific
Supplier: Globe Scientific
456106 456105
89498-000EA 34.16 USD InStock
89498-000 89498-002
Test Tube Racks, Globe Scientific
Racks Tube Racks
These traditional style test tube holders are lightweight, compact, and unbreakable
Test tube racks are manufactured from durable polypropylene and can be autoclaved at 121°C for 20 minutes.
They take up minimal space on the benchtop to provide a larger work area.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty