Would you like to log in again?
Stainless Steel Racks for ¹²/₁₃ mm Tubes, Globe Scientific
Supplier: Globe Scientific
457100 457105
89498-066EA 42.37 USD InStock
89498-066 89498-064
Stainless Steel Racks for ¹²/₁₃ mm Tubes, Globe Scientific
Racks Tube Racks
Specially shaped 'Z' racks are rust-proof and will hold up to repeated use.
  • 25-place rack (5×5 format) dimensions: 4.5×4.25×1.5"
  • 50-place rack (5×10 format) dimensions: 8.75×4.5×2"
Stainless steel racks are autoclavable.
The wide base provides stability and prevents tipping. When not in use, racks stack neatly inside one another to conserve benchtop space.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty