Would you like to log in again?
Aluminum Crimp Seals with Liner, 20 mm, Ace Glass Incorporated
Supplier: Ace Glass
5705-10
89427-790CS 1203.4 USD InStock
89427-790
Aluminum Crimp Seals with Liner, 20 mm, Ace Glass Incorporated
Closures Chromatography Vial Closures
20mm Aluminum crimp seals for Microwave reaction vials
Open top with 0.125-in thick PTFE faced silicone liner.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty