Would you like to log in again?
Moisture Test Receiver, Recycle Type, Ace Glass Incorporated
Supplier: Ace Glass
7747-19 7747-04 7747-17 7747-02
89060-356EA 776.73 USD InStock
89060-356 89060-358 89060-360 89060-362
Moisture Test Receiver, Recycle Type, Ace Glass Incorporated
Distillation Components Distillation Receivers
Especially useful when heavier-than-water solvents are employed
A 120°, 1:5 PTFE stop-cock provides adequate shut-off in the intermediate position. Thermometer joint is a standard taper 10/30.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty