Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diboron trioxide, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Boron oxideBoric anhydride

1.00169.0250 1.00169.0250
EM1.00169.0250CS 38203 USD InStock
CA1.00169.0250 EM1.00169.0250
Diboron trioxide, Suprapur® for trace metal analysis, Supelco®
di-Boron trioxide


Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: 1860 °C (1013 hPa)
Melting Pt: 450 °C
Density: 1.81…1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 12,01365
Order Now
Specification Test Results
Solubility 36 g/l (25 °C)
Melting Point 450 °C
Molar Mass 69.61 g/mol
Bulk Density 900 - 1100 kg/m3
Boiling Point 1860 °C
Density 2.46 g/cm3 (20 °C)
pH 4 (10 g/l, H2O, 25 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty