Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dodecamolybdophosphoric acid solution in 2-propanol, spray reagent for TLC, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Phosphomolybdic acidMolybdophosphoric acid

M-Clarity™ quality level = MQ100

1.00480.0100 1.00480.0100
EM1.00480.0100EA 110.79 USD InStock
CA1.00480.0100 EM1.00480.0100
Dodecamolybdophosphoric acid solution in 2-propanol, spray reagent for TLC, Supelco®
Dodecamolybdophosphoric acid
Ready-to-use spray solution.


Formula: H₃Mo₁₂O₄₀P
MW: 1825.25 g/mol
MDL Number: MFCD00011341
CAS Number: 12026-57-2
UN: 1219
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty