Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid ≥96%, Ultrapur for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.01516.0250 1.01516.0250
EM1.01516.0250CS 30904 USD InStock
CA1.01516.0250 EM1.01516.0250
Sulfuric acid ≥96%, Ultrapur for trace metal analysis, Supelco®
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974
Order Now
Specification Test Results
Solubility (20 °C) soluble,(caution ! development of heat)
Melting Point -20 °C
Molar Mass 98.08 g/mol
Boiling Point 335 °C
Vapor Pressure 0.0001 hPa (20 °C)
Density 1.84 g/cm3 (20 °C)
pH 0.3 (49 g/l, H2O, 25 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty