Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium hydroxide octahydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.01737.0500 1.01737.0500
EM1.01737.0500CS 1406 USD InStock
EM1.01737.0500 CA1.01737.0500
Barium hydroxide octahydrate, EMSURE® for analysis, Supelco®
Barium hydroxide octahydrate


Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Melting Pt: 505 °C
Density: 3.2 g/cm³ (20 °C)
Flash Pt: 21 °C (closed cup)
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 236-710-4
UN: 2923
ADR: 8 (6.1),II
Merck Index: 12,01016
Order Now
Specification Test Results
Solubility 72 g/l (20 °C)
Melting Point 78 °C
Molar Mass 315.48 g/mol
Bulk Density 900 - 1100 kg/m3
Density 2.18 g/cm3 (20 °C)
pH 12.5 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty