Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium fluoride, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06450.0025 1.06450.0025
EM1.06450.0025CS 36954 USD InStock
EM1.06450.0025 CA1.06450.0025
Sodium fluoride, Suprapur® for trace metal analysis, Supelco®
Sodium fluoride


Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 12,08762
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/30/16640630.jpg
Order Now
Specification Test Results
Solubility 42 g/l (20 °C)
Melting Point 993 °C
Molar Mass 41.98 g/mol
Bulk Density 800 kg/m3
Boiling Point 1704 °C
Vapor Pressure 1 hPa (1077 °C)
Density 2.8 g/cm3 (20 °C)
pH 10.2 (40 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty