Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PSS (Phosphoric acid, Sulfuric acid), Spectrosolv® acid blank, for the ACIDORAPID® mineralizer, Supelco®

Supplier: MilliporeSigma
Danger

1.07916.1000 1.07916.1000
EM1.07916.1000CS 14025 USD InStock
EM1.07916.1000 CA1.07916.1000
PSS (Phosphoric acid, Sulfuric acid), Spectrosolv® acid blank, for the ACIDORAPID® mineralizer, Supelco®
PSS (Phosphoric acid, Sulfuric acid)


Formula: H5O8PS
MW: 196,08 g/mol
UN: 3264
ADR: 8,II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Density 1.78 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty