Would you like to log in again?
OPTI-LYNX™ Hardware - Individual Pieces
Supplier: Optimize Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
11-02212-AA
77100-302EA 312.08 USD InStock
77100-302
OPTI-LYNX™ Hardware - Individual Pieces
Chromatography Supplies Chromatography Spare Parts
Opti-Lynx inserts can be a filter, guard column, trap cartridge or analytical column.
Since the interacting surfaces are identical, the only variable component is the holder tube. Install a guard column for one application and then switch to a pre-column filter for the next. Use an in-line filter with different porosities and diameters just by changing the insert. Switch out columns in an instant with the Velocity® system or retrofit current columns with the universal port adapter. You can make and break flow paths for multiple detectors, or anywhere else a break in flow is frequently made. Opti-Lynx allows all of this without ever having to change out the basic components.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty