Would you like to log in again?
Distillation Head with Plugs, Vacuum Type, Ace Glass Incorporated
Supplier: Ace Glass
7792-20 7792-15 7792-06 7792-04 7792-07
89060-416EA 1140.29 USD InStock
89060-416 89060-418 89060-422 89060-424 89060-420
Distillation Head with Plugs, Vacuum Type, Ace Glass Incorporated
Distillation Components Distillation Heads
Used for distilling solvents directly into Schlenk tubes
With standard taper 10/18 thermometer joint for a 51mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty