Would you like to log in again?
Buffer, Phosphate Type, pH 7, for Amperometric Chlorine, 100 ml MDB, Hach
Supplier: Hach
2155332
97026-638EA 50.52 USD InStock
97026-638
Buffer, Phosphate Type, pH 7, for Amperometric Chlorine, 100 ml MDB, Hach
Environmental Test Kits
For determination of chlorine by amperometric titration
APHA formulation 100 mL bottle.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty