Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiosulfate 2.26 N stabilized, cartridge for Digital Titrator

Synonyms: di-Sodium thiosulfateSodium hyposulphitedi-Sodium thiosulphateSodiumthiosulfateSodium hyposulfite

2686901 2686901
97026-356EA 57.72 USD InStock
97026-356 CA11016-468
Sodium thiosulfate 2.26 N stabilized, cartridge for Digital Titrator
Sodium thiosulphate
For determination of Hypochlorite (Bleach) by Digital Titrator titration.

  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical

Hach Method 10100. Range: 5 - 15% Cl2 (50 - 150 g/L Cl2). Stabilized sodium thiosulfate formulation. 2.26 N Digits x 0.5 = g/L CI2 in 0.2 mL sample.


Formula: Na₂S₂O₃
MW: 158.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Order Now
Specification Test Results
Method Name Iodometric Method
Number of tests 40 - 100
Parameter Hypochlorite
Platform Digital Titrator
Range 50 - 150 g/L Cl2
Titrant 2.26 N Na2S2O3

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty