Would you like to log in again?
SP Bel-Art Full-View Test Tube Supports, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H18914-0000 H18910-0102 H18911-0000 H18912-0000 H18915-0000 H18910-0189
60917-606EA 59.25 USD InStock
60917-606 60917-609 60917-603 60917-615 60917-600 60917-612
SP Bel-Art Full-View Test Tube Supports, Bel-Art Products, a part of SP
Racks Tube Racks
This versatile polypropylene rack has pins for supporting test tubes upright, full or empty, or holding inverted test tubes for draining.
  • Polypropylene construction
  • Autoclavable
  • Versatile
Rack 60917-600, with 102 pins, 5 x 16 rows, is excellent for drying and storing glass plates used with vertical electrophoresis apparatus. Steam autoclavable at 121°C (250°F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty