Would you like to log in again?
Human A1-Acid Glycoprotein (from Plasma)
Supplier: EMD Millipore
112251-1MG 112251-1MG
80055-620EA 169.63 USD InStock
80055-620 CA80055-620
Human A1-Acid Glycoprotein (from Plasma)
Proteins and Peptides
Native α1-acid glycoprotein from human plasma. Glycoprotein used for the study of the structure of oligosaccharide units. Consists of 40 - 45% carbohydrate. Present in human plasma at concentrations of 55 - 140 mg/100 mL. Clinically, α1-AG is an acute-phase reactant that, together with haptoglobin, indicates acute inflammation. Selectively suppresses the augmentation of NK cell activity by IFN-α and IFN-ɣ.
Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty