Would you like to log in again?
Chlorine (Free and Total) Reagent Set, DPD FEAS, Digital Titrator, Hach
Supplier: Hach
2445300
97027-056EA 133.4 USD InStock
97027-056
Chlorine (Free and Total) Reagent Set, DPD FEAS, Digital Titrator, Hach
Environmental Test Kits
  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical
For determination of Chlorine (Free and Total) by Ferrous Ethylenediammonium Sulfate Digital Titration, using DPD indicator. Range: Up to 3.00 mg/L Cl2. Reagent Set includes: DPD Free Chlorine Reagent Powder Pillows, DPD Total Chlorine Reagent Powder Pillows, Ferrous Ethylenediammonium Sulfate Titration Cartridge, 0.00564 N. Approximately 40-100 tests. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty