Would you like to log in again?
Universal-Fitting MicroFlex® Gel-Loading Pipette Tips, National Scientific Supply
Supplier: National Scientific
Microflex®
MN120U-LR MN320F-LRS MN520R-LR MN320F-LR MN520R-LRS
53503-167CS 317.7 USD InStock
53503-167 53503-189 53503-178 53503-193 53503-190
Universal-Fitting MicroFlex® Gel-Loading Pipette Tips, National Scientific Supply
Pipette Tips
MicroFlex® Gel-loading pipette tips are universal-fitting polypropylene pipette tips.
The tips are individually wrapped, Rnase and Dnase-free. Tips are not autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty