Would you like to log in again?
Model CA-10WR Carbon dioxide, Dissolved Oxygen and pH Test Kit, Hach
Supplier: Hach
143801 143801
89500-640EA 617.65 USD InStock
CA89500-640 89500-640
Model CA-10WR Carbon dioxide, Dissolved Oxygen and pH Test Kit, Hach
Environmental Test Kits
Water ecology and limnology pertain to the physical, chemical and biological properties of fresh water, especially lakes, ponds and streams
  • Uses simple, fast color disc method to measure carbon dioxide, dissolved oxygen, and pH
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
These properties are essential for maintaining water conditions that are capable of supporting aquatic life. Many of these properties require that the measurement be made directly on-site.
Ordering information: Seven reagents, clippers, bottles, sample cell with cap, color disc, viewing tubes, comparator box, measuring tube, instructions and carrying case included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty