Would you like to log in again?
Polyethylene Hollow Stopper, Size 1, Hach
Supplier: Hach
1448000 1448000
89500-642PK 38.45 USD InStock
CA89500-642 89500-642
Polyethylene Hollow Stopper, Size 1, Hach
Stoppers PE/LDPE/HDPE Stoppers
Made of low-density polyethylene is interchangeable with rubber stoppers
Safer and more economical than glass, the stopper fits snugly. Excellent as sample cup for pH meters, flame photometers and automatic analyzers, or as a small beaker. 25-26 mm/ 1" high.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty