Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plastic Dropper for 59 ml Bottle, Hach

Supplier: Hach
2355237
89500-726EA 10.07 USD InStock
89500-726
Plastic Dropper for 59 ml Bottle, Hach
Pipettes Pasteur/Transfer Pipettes
Child-resistant dropper assembly with 0.5 and 1.0 ml marks on plastic barrel.

Fits 59 ml bottle.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty