Would you like to log in again?
SP Bel-Art Sample Vials with Captive Closure, LDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F17570-0000 F17574-0000 F17573-0000
70680-372CS 500.05 USD InStock
70680-372 66001-102 70680-376
SP Bel-Art Sample Vials with Captive Closure, LDPE, Bel-Art Products, a part of SP
Vials
Cylindrical, low-density PE vials with friction-fit, captive closures open and close with a flick of your thumb.
  • LDPE construction
  • Captive closures controlled with thumb pressure
  • Suited for storing liquids or storing and shipping dry materials
Excellent for storage of liquids as well as for storing and shipping dry materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty