Would you like to log in again?
Advanced Digital Free Fall Timer and Stopwatch
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SV689
470211-168CS 1219.8 USD InStock
470211-168
Advanced Digital Free Fall Timer and Stopwatch
Educational Materials Physics Educational Materials Mechanics Learning Activities
ThIs advanced digital free fall timer and stopwatch uses modern electronics, piezo landing pad, and a push button launcher to accurately and conveniently perform free-fall experiments with fall distance of 50 cm to over 2m. Features include a large LED display, data storage up to five entries, a landing pad, push button, single on/off switch. Memory status, and sensor pulse and ball status shown by LEDs and built-in stopwatch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty