Would you like to log in again?
Bowie-Dick Type Air Removal Test Packs, Mesa Laboratories
Supplier: Mesa Laboratories
77-05CI-BDTLF20
101384-633PK 207.03 USD InStock
101384-633
Bowie-Dick Type Air Removal Test Packs, Mesa Laboratories
Sterilization Indicators
Air Removal Test Packs are intended to evaluate the performance of the air removal (vacuum) system of pre-vacuum equipped steam sterilizers.
Pre-assembled, disposable test pack. After exposure, unwrap pack and remove the indicator sheet for your records. Lead-free ink used on the indicator sheet is safer for medical personnel and the environment. Distinct, non-reversible pink to black color change makes results easy to interpret.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty