Would you like to log in again?
Peptone Water, Hardy Diagnostics
Supplier: Hardy Diagnostics
K47 K47
89423-878PK 88.22 USD InStock
CA89423-878 89423-878
Peptone Water, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Peptone Water, 9ml, 20x125mm tube, order by the package of 20, by Hardy Diagnostics. MINIMUM ORDER: 5 packs.
For use as a diluent and for the preparation of suspensions. Contains 0.1% proteose peptone and 1% NaCl.
Peptone Water, 9ml, 20x125mm tube, order by the package of 20, by Hardy Diagnostics
MINIMUM ORDER: 5 packs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty