Would you like to log in again?
Fluid D, Hardy Diagnostics
Supplier: Hardy Diagnostics
U346 U346
89498-956PK 815.51 USD InStock
89498-956 CA89498-956
Fluid D, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Fluid D, USP, 300ml fill, 500ml glass bottle with needle-port septum screw cap, order by the package of 10, by Hardy Diagnostics.
USP Fluids A, D and K are used by the pharmaceutical and medical device industry to insure sterility of injectables, non-injectables, liquid products, oils and ointments, antibiotic powders, sterile pathway devices and aerosols.
Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.
Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center
Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.
Certifications: USP
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty