Would you like to log in again?
Letheen Broth with Tween, Hardy Diagnostics
Supplier: Hardy Diagnostics
U355 U355
89498-958PK 666.98 USD InStock
CA89498-958 89498-958
Letheen Broth with Tween, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Letheen Broth, Modified with Tween 80 1.5%, 1000ml bottle, 1000ml fill, order by the package of 10, by Hardy Diagnostics
For sample preparation and dilution for cosmetic and personal care product samples. Use in determining the antimicrobial activity of quaternary ammonium compounds and in the microbial evaluation of cosmetics.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty