Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
OxiDrops™ Liquid Oxidase Reagent, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
OxiDrops™
Z119 Z119
89428-918EA 60.37 USD InStock
89428-918 CA89428-918
OxiDrops™ Liquid Oxidase Reagent, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
OxiDrops™, liquid oxidase reagent for spot test, 7ml, dropper bottle, by Hardy Diagnostics. This product has a short shelf life and ships from our California warehouse only.
For testing the oxidase reaction, contains tetramethyl-p-phenylenediamine. Simply place one drop on the suspect colony (directly on the plate) and look for the color reaction.
OxiDrops™, liquid oxidase reagent for spot test, 7ml, dropper bottle, by Hardy Diagnostics
This product has a short shelf life and ships from our California warehouse only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty