Would you like to log in again?
Potassium Hydroxide (KOH), Hardy Diagnostics
Supplier: Hardy Diagnostics
Z173
89428-920EA 61.86 USD InStock
89428-920
Potassium Hydroxide (KOH), Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Potassium Hydroxide (KOH), for fungal skin prep, 3%, 15ml, dropper vial, order by the each, by Hardy Diagnostics.
For use in slide preparations of specimens for the purpose of identifying fungal elements.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty