Would you like to log in again?
Indole, DMACA Spot, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z65 Z65
89428-932EA 28.92 USD InStock
89428-932 CA89428-932
Indole, DMACA Spot, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Indole, DMACA Spot, for indole spot test on filter paper, cinnamaldehyde, 15ml dropper vial, by Hardy Diagnostics.
Not for the tube indole test.
Ordering information: See Kovac's Indole for tube indole test, catalog number 89407-610 .
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty