Would you like to log in again?
Adhesive Labels, Round, USA Scientific
Supplier: USA Scientific
Tough-Spots®
9185-2004 9185-5000 9185-2002 9185-0500 9185-3000 9185-1004 9185-0500 9185-1006 9185-4000 9185-2000 9185-2001 9185-2003 9185-2006 9185-1008 9185-1000 9185-1003 9185-2008
102953-828CS 97.92 USD InStock
102953-828 102953-826 102952-568 102953-834 102952-702 102953-824 82020-736 102952-560 102953-362 102953-832 102953-546 102953-822 102953-830 102953-820 102953-982 102953-840 102953-992
Adhesive Labels, Round, USA Scientific
Labels
Tough-Spots® Labels are chemically resistant, flexible labels that can withstand boiling, freezing to −196 °C, sustained humidity.
  • Polyolefin labels resist most organic solvents and caustic agents
  • Label sheets are specially top-coated to readily accept laser printing
  • Assortment includes blue, green, orange, red and yellow
These labels fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers. They adhere to most plastics, glass and metals without cracking, peeling, or degrading.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty