Would you like to log in again?
SP Bel-Art Large Storage Bin, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SCIENCEWARE®
F18669-0000
47750-966EA 247.16 USD InStock
47750-966
SP Bel-Art Large Storage Bin, Bel-Art Products, a part of SP
Boxes Dispenser Boxes
Keep small items organized and within reach.
  • Clear front panel
  • White acrylic construction
  • Removable top
  • Dust-free storage
Constructed of white acrylic, this large storage bin has a clear acrylic front panel that keeps contents in view. Ideal for dust-free storage of bulk purchase items such as tubes, tips and cotton balls. The top is removable for filling and the hinged cover allows easy dispensing of items as needed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty