Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ampicillin sodium salt

Supplier: GROWCELLS MS
Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

MBPC-1900
101384-974EA 162.14 USD InStock
101384-974
Ampicillin sodium salt
Ampicillin sodium salt
A beta-lactam broad spectrum antibiotic used to select ampicillin-resistant plasmids

Ampicillin is a penicillin derivative that inhibits bacterial cell-wall synthesis and is inactivated by lactamases

Certifications: USP Grade


Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty