Would you like to log in again?
X-gal/IPTG solution 8mg/ml, FIROZEH, non toxic ready-to-use
Supplier: GROWCELLS MS
Synonyms: X-GAL & IPTG
MESL-3001 MESL-3002
101385-628EA 149.94 USD InStock
101385-628 101385-630
X-gal/IPTG solution 8mg/ml, FIROZEH, non toxic ready-to-use
X-Gal/IPTG
Ready to use X-Gal/IPTG solution designed for selection of cloning construct in LacZ expression in the presence of lacl repressor
  • 8mg/mL each of X-Gal and IPTG
Stable at 4C reducing freeze-thaw cycles that reduce selection efficiency. Produced from the non-toxic ingredients, eliminating the handling of hazardous DMSO or DMF
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty