Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiosulfate pentahydrate 99.5-101.0%, crystals ACS

Supplier: VWR

ANAC85778-300
CA71008-848EA 94.54 USD InStock
CA71008-848
Sodium thiosulfate pentahydrate 99.5-101.0%, crystals ACS
Sodium thiosulphate pentahydrate


Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844
Order Now
Specification Test Results
Assay (Na2S2O3.5H2O) 99.5 - 101.0%
pH of a 5% Solution at 25°C 6.0 - 8.4
Insoluble Matter 0.005% max.
Nitrogen compounds (as N) 0.002% max.
Sulfate and sulfite (as SO4) 0.1% max.
Sulfide (S) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty