Would you like to log in again?
SP Wilmad-LabGlass Dewar Shape Vacuum Traps, SP Industries
Supplier: SP Industries
LG-11065-094
14205-652EA 1651.83 USD InStock
14205-652
SP Wilmad-LabGlass Dewar Shape Vacuum Traps, SP Industries
Vacuum Traps
Designed to remove relatively large volumes of solvent, these borosilicate glass vacuum traps can be used with rotary evaporators
Inner cylinder is charged with dry ice/alcohol or liquid nitrogen. Lower reservoir has a PTFE valve to remove condensate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty