Would you like to log in again?
96-Well Microtest Plates, Sarstedt
Supplier: SARSTEDT INC MS
82.1581.001 82.1583 82.1583.001
101093-338CS 127.19 USD InStock
101093-338 103574-400 101093-365
96-Well Microtest Plates, Sarstedt
Microplates HTS Microplates
The 96-Well Microtest plates are compatible for use with all common dispensers, readers, and washers.
They provide a range of applications, including blood group serology, haemagglutination tests, antibody diagnostic tests, and complement binding reactions. Polystyrene plates are available with flat, round, or conical bottoms.
Ordering information: Lids must be ordered separately.
Packaging: Sterile plates are individually wrapped with lids, and nonsterile plates without lids are packaged in bags of 25.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty