Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3-chloropyridine ≥98% (by HPLC), Powder

Supplier: CHEM IMPEX INTL INC MS

17023-25G
102639-720EA 89.14 USD InStock
102639-720
2-Amino-3-chloropyridine ≥98% (by HPLC), Powder
2-Amino-3-chloropyridine


Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD03541052
CAS Number: 39620-04-7
EINECS: 000-000-0
Order Now
Specification Test Results
Molecular Formula C5H5ClN2
Molecular Weight 128.56
Melting Point 59-65 ºC
MDL Number MFCD03541052
Storage Temperature Store at 0-8°C
Color Gray to Yellow

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty