Would you like to log in again?
Murashige & Skoog Medium, HiMedia Laboratories
Supplier: HiMedia Laboratories
PT049-25L PT049-5L PT049-5L PT049-25L
95025-924EA 23.53 USD InStock
CA95025-926 95025-924 CA95025-924 95025-926
Murashige & Skoog Medium, HiMedia Laboratories
Cell Culture Media
The Murashige & Skoog Medium has been specially formulated for plant cell, tissue and organ cultures
Dehydrated plant tissue culture media powder is extremely hygroscopic and should be protected from atmospheric moisture. If possible, the entire content of each bottle should be used immediately after opening or else the unused portion should be stored in a desiccator and refrigerated at 2 to 8 °C. Use before the expiry date.
Potassium nitrate serves as the nitrate source. Glycine serves as the source of amino acid. Medium does not contain sucrose and agar; hence these components have to be added to the medium before use. With vitamins, without NH4NO3, Sucrose, and Agar. Light yellow to yellow, homogeneous, free flowing powder appearance. 2.76 gm/litre soluble in distilled water solubility. 4.0 ±0.5 of 0.276% w/v dehydrated medium pH at 25 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty