Would you like to log in again?
Filter Reactor Head, Ace Glass Incorporated
Supplier: Ace Glass
6527-26
89055-318EA 605.19 USD InStock
89055-318
Filter Reactor Head, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
This reactor head has a 60 mm O-ring groove flange with 3 in-line 24/40 joints
Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty