Would you like to log in again?
Canister HEPA Vacuum, Atrix
Supplier: Atrix
AHC-1
100720-990EA 227.31 USD InStock
100720-990
Canister HEPA Vacuum, Atrix
Vacuum Cleaners
With its 3 stage HEPA filtration, the Atrix Canister Vacuum captures and removes fine particles such as dust and pet dander quickly and efficiently
The variable speed motor allows the Canister Vac to adapt to any cleaning situation. The Atrix Canister Vacuum features a full-bag indicator light and is light-weight and easy to pull.
Ordering information: Includes 2 HEPA Bags, 20' retractable power cord, 6' hose, meal extension wand, turbo brush, floor brush, crevice tool with brush, oval dusting brush and upholstery nozzle
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty