Would you like to log in again?
Filter Stick, Ace Glass Incorporated
Supplier: Ace Glass
15337-09 15337-05 15337-07 15337-03
89078-326PK 195.11 USD InStock
89078-326 89078-324 89078-328 89078-322
Filter Stick, Ace Glass Incorporated
Filters Filter Tubes
This is a porous fritted glass filter stick used to fabricate gas dispersion tubes.
Hollow at one end.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty