Would you like to log in again?
Cogent Phenyl Hydride™ TYPE-C Silica™ HPLC Columns, (100 Å Pore Size), MicroSolv
Supplier: MicroSolv
69020-25P-1 69020-75P 69020-05P-3
89218-906EA 1007.6 USD InStock
89218-906 89218-926 89218-932
Cogent Phenyl Hydride™ TYPE-C Silica™ HPLC Columns, (100 Å Pore Size), MicroSolv
Chromatography Columns
These useful HPLC columns have a silicon hydride surface similar to other TYPE-C™ phases but the uniqueness of Phenyl Hydride™ is that they have unique selectivity for compounds with aromatic rings with all the TYPE-C silica benefits.
These columns can be used in RP, ANP or NP and changed back and forth without damage to the columns.
Ordering information: Guard column kits contain five guard columns with or without a holder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty