Would you like to log in again?
High Capacity HEPA Vac, Atrix
Supplier: Atrix
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
ATIHCTV5CT
101640-732EA 707.34 USD InStock
101640-732
High Capacity HEPA Vac, Atrix
Vacuum Cleaners
The High Capacity 5 Gallon HEPA dry particulate vacuum is an excellent choice where high volumes of non-hazardous or hazardous dry particulate needs to vacuumed in or around static sensitve machines or components
HEPA Efficiency: 99.97% @ .3 microns. Vacuum components are all economical and ESD Safe.
Ordering information: Includes one HP flow-through motor, high capacity ESD safe toner vacuum, one super-flex, ESD aafe toner-proof hose, ESD safe crevice tool with brush, 16' ESD safe rubber flex-neck tool, detachable power cord, roller base, ESD safe 5 gallon HEPA filter bucket
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty