Would you like to log in again?
Backpack HEPA Vac and Blower, Atrix
Supplier: Atrix
VACBP1
100720-984EA 260.11 USD InStock
100720-984
Backpack HEPA Vac and Blower, Atrix
Vacuum Cleaners
The Atrix Backpack HEPA Vacuum/Blower with an 8 Quart dry particulate filter is ideal for any situation that involves a lot of cleaning, stairs, draperies, blinds, etc.
  • Ergonomic and comfortable fit for all sizes
  • Weighs only 10 pounds
  • HEPA filter designed to capture 99.97% of particles as small as 0.3 microns
Ordering information: Includes hose, adjustable extension wand, 1.5' power cord, hard floor tool, BP44 Floor Turbo brush, oval brush, furniture brush and microattachment kit.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty