Would you like to log in again?
Omega Supreme Laboratory ULPA Vac, Atrix
Supplier: Atrix
VACOMEGASLFU
100720-968EA 410.26 USD InStock
100720-968
Omega Supreme Laboratory ULPA Vac, Atrix
Vacuum Cleaners
The Omega Supreme Lab Plus Vacuum is ESD safe, quiet, powerful, and the new motor uses less energy than standard vacuums
Replacement Filters are designed to capture fumes, atmospheric dust, black toner and all color toner and other ultra-fine particulate. Compatible with 3M vacuums.
This vacuum features an electronic line filter that supresses EMI/RFI line noise. It includes immediate containment, critical-application ULPA cartridge for bacteria, asbestos, mold, lead dust, arsenic, and other small hazardous particles. The Omega Supreme Lab Plus Vacuum also features over-heat protection and field replaceable latches.
Ordering information: Includes quiet 1HP flow-thru motor with sound baffles, self-storing utensils (cover insert), ESD safe crevice tool with brush, 16" ESD safe rubber flex-neck tool, detachable power cord, shoulder strap with D-rings, HEPA filter, fully statically controlled
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty