Would you like to log in again?
Omega Supreme MRI / Hospital HEPA Vac, Atrix
Supplier: Atrix
VACOSMRI
101640-736EA 493.34 USD InStock
101640-736
Omega Supreme MRI / Hospital HEPA Vac, Atrix
Vacuum Cleaners
The Omega Supreme MRI 1 Gallon HEPA dry particulate vacuum with 15' Non-Metallic Hose is perfect for the maintenance of MRI machines
Replacement Filters are designed to capture fumes, atmospheric dust, black toner and all color toner and other ultra-fine particulate. Compatible with 3M vacuums.
With the standard 15' hose, service personnel can keep the vacuum a safe distance away from the magnetic field. ULPA filter option for an even higher level of containment. HEPA Efficiency: 99.97% @ 0.3 microns.
Ordering information: Includes quiet 1HP flow-thru motor with sound baffles, self-storing utensils (cover insert), 15' ESD non-metallic hose, ESD crevice tool, 16" ESD gooseneck, shoulder strap with D-Rings, HEPA filter, electronic line filter, 10' power cord, fully statically controlled
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty