Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Coenzyme A trilithium salt, Sigma-Aldrich®

Supplier: MilliporeSigma

234101-100MG 234101-100MG
80051-106EA 289.75 USD InStock
CA80051-106 80051-106
Coenzyme A trilithium salt, Sigma-Aldrich®
Coenzyme A trilithium salt
A cofactor in enzymatic acetyl transfer reactions. Hygrocopic.


Formula: C₂₁H₃₃N₇O₁₆P₃SLi₃
MDL Number: MFCD00075848
CAS Number: 18439-24-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty