Would you like to log in again?
IntelliCAL™ PHC281 Ultra Refillable pH Electrode, Hach
Supplier: Hach
PHC28103 PHC28103
89501-174EA 1293.8 USD InStock
89501-174 CA11014-074
IntelliCAL™ PHC281 Ultra Refillable pH Electrode, Hach
Electrodes pH Electrodes
IntelliCAL™ PHC281 pH Ultra probe is a standard, digital, combination pH probe with a refillable double junction reference and built-in temperature sensor
The PHC281 pH Ultra probe is designed for premium performance, even in difficult samples. It provides ultimate traceability in measurement history, and the user is alerted when re-calibration is needed. IntelliCAL™ digital pH probe can be moved between meters without the need to re-calibrate or re-enter measurement settings.
It is available with a 1 or 3 meter cable and is intended for laboratory use. The PHC281 pH probe is designed for fast response and stabilization. It is ideal for measuring pH in general purpose water quality applications or in difficult samples such as low ionic strength (LIS), ultrapure, dirty, TRIS, or high-solid content samples.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty