Would you like to log in again?
IntelliCAL™ Fluoride ISE Analysis Package, Hach
Supplier: Hach
ISEF121AP ISEF121AP
89501-106EA 2396.69 USD InStock
CA89501-106 89501-106
IntelliCAL™ Fluoride ISE Analysis Package, Hach
Electrodes ISE Electrodes
The IntelliCAL™ Fluoride ISE Analysis Package contains the IntelliCAL™ ISEF121 Fluoride ISE, 100 pack of fluoride ISA powder pillows, 500 ml of 0.5 mg/L fluoride standard solution, 500 ml of 1.0 mg/L fluoride standard solution, and 500 ml of 2.0 mg/L fluoride standard solution.
This analysis package requires virtually no maintenance, and it is fast and stable. Results are accurate and have ultimate traceability in measurement history. Componants can be moved between meters without the need to re-calibrate or re-enter measurement settings. Thermistor is integrated. Pack requires dry storage. No replacement membranes.
IntelliCAL™ ISEF121 is a digital, combination fluoride ion selective electrode (ISE) with a non-refillable, single junction reference and built-in temperature sensor. The IntelliCAL ISEF121 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISEF121 is ideal for measuring fluoride concentrations in drinking water, wastewater, and general water quality applications. The ISEF121’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty